siRNA Details
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290504.1 | NP_001277433.1 | 1169 | |||
NM_001290505.1 | NP_001277434.1 | 1393 | ||||
NM_001290506.1 | NP_001277435.1 | 1279 | ||||
NM_001290512.1 | NP_001277441.1 | 613 | ||||
NM_010797.3 | NP_034927.2 | 1276 | ||||
GenBank mRNA | AF026565.1 | - | 1169 | |||
AK160120.1 | - | 615 | ||||
BC053704.1 | - | 1246 | 30058736 | |||
BC059027.1 | - | 270 | 6810837 | |||
Y14848.1 | - | 1230 |
