siRNA Details
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290504.1 | NP_001277433.1 | 1251 | |||
NM_001290505.1 | NP_001277434.1 | 1475 | ||||
NM_001290506.1 | NP_001277435.1 | 1361 | ||||
NM_001290512.1 | NP_001277441.1 | 695 | ||||
NM_010797.3 | NP_034927.2 | 1358 | ||||
GenBank mRNA | BC053704.1 | - | 1328 | 30058736 | ||
BC059027.1 | - | 352 | 6810837 | |||
Y14848.1 | - | 1312 |
