siRNA Details
| Gene Symbol: Mip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008600.4 | NP_032626.2 | 2 | 483 | ||
| GenBank mRNA | AK053490.1 | - | 2 | 484 | ||
| AK053494.1 | - | 2 | 446 | |||
| AK136287.1 | - | 2 | 484 | |||
| BC082567.1 | - | 2 | 472 | 30551292 | ||
| U27502.1 | - | 2 | 439 | |||

Mip
Genomic Map