siRNA Details
| Gene Symbol: Bhlha15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010800.4 | NP_034930.1 | 2 | 487 | ||
| GenBank mRNA | AF049660.1 | - | 2 | 484 | ||
| AK020643.1 | - | 2 | 446 | |||
| BC011486.1 | - | 1 | 437 | 4189225 | ||
| BC094061.1 | - | 2 | 463 | 4165423 | ||

Bhlha15
Genomic Map