siRNA Details
| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081117.2 | NP_001074586.2 | 6 | 899 | ||
| GenBank mRNA | AK052772.1 | - | 6 | 819 | ||
| AK082099.1 | - | 6 | 817 | |||
| AK143677.1 | - | 6 | 938 | |||
| X82786.1 | - | 6 | 1007 | |||

Mki67
Genomic Map