siRNA Details
| Gene Symbol: Mknk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285487.1 | NP_001272416.1 | 13 | 1215 | ||
| NM_001285488.1 | NP_001272417.1 | 12 | 1109 | |||
| NM_021461.5 | NP_067436.1 | 11 | 1065 | |||
| GenBank mRNA | AK141298.1 | - | 9 | 2923 | ||
| AK155934.1 | - | 13 | 1176 | |||
| AK169982.1 | - | 12 | 1075 | |||
| BC021369.1 | - | 11 | 1021 | 5030044 | ||
| CN538557.1 | - | 3 | 333 | |||
| Y11091.1 | - | 11 | 1030 | |||

Mknk1
Genomic Map