siRNA Details
| Gene Symbol: Mknk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285487.1 | NP_001272416.1 | 6, 7 | 578 | ||
| NM_001285488.1 | NP_001272417.1 | 5, 6 | 472 | |||
| NM_021461.5 | NP_067436.1 | 4, 5 | 428 | |||
| GenBank mRNA | AK010697.1 | - | 4, 5 | 406 | ||
| AK141298.1 | - | 4, 5 | 406 | |||
| AK155934.1 | - | 6, 7 | 539 | |||
| AK169982.1 | - | 5, 6 | 438 | |||
| BC021369.1 | - | 4, 5 | 384 | 5030044 | ||
| BQ551007.1 | - | 2, 3 | 135 | |||
| Y11091.1 | - | 4, 5 | 393 | |||

Mknk1
Genomic Map