siRNA Details
| Gene Symbol: Mme | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289462.1 | NP_001276391.1 | 6 | 921 | ||
| NM_001289463.1 | NP_001276392.1 | 6 | 705 | |||
| NM_008604.4 | NP_032630.2 | 6 | 555 | |||
| GenBank mRNA | AK031446.1 | - | 6 | 556 | ||
| AK033824.1 | - | 6 | 616 | |||
| AK040730.1 | - | 6 | 592 | |||
| AK052088.1 | - | 5 | 442 | |||
| AK076908.1 | - | 6 | 769 | |||
| AK080202.1 | - | 6 | 583 | |||
| BC034092.1 | - | 6 | 577 | 4483558 | ||
| BC066840.1 | - | 5 | 464 | |||
| M81591.1 | - | 5 | 467 | |||

Mme
Genomic Map