siRNA Details
| Gene Symbol: Mmp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008607.2 | NP_032633.1 | 7 | 1004 | ||
| GenBank mRNA | AK150728.1 | - | 7 | 991 | ||
| AK151626.1 | - | 7 | 991 | |||
| AK152507.1 | - | 7 | 990 | |||
| AK152916.1 | - | 7 | 991 | |||
| AK153458.1 | - | 7 | 989 | |||
| AK155188.1 | - | 7 | 990 | |||
| AK159921.1 | - | 7 | 991 | |||
| AK160819.1 | - | 7 | 989 | |||
| AY622973.1 | - | 1 | 44 | |||
| BC125320.1 | - | 7 | 1004 | 40129835 | ||
| BC125322.1 | - | 7 | 1004 | 40129837 | ||
| X66473.1 | - | 7 | 973 | |||

Mmp13
Genomic Map