siRNA Details
| Gene Symbol: Ndst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010811.2 | NP_034941.2 | 3 | 1682 | ||
| GenBank mRNA | AF074925.1 | - | 1 | 932 | ||
| AK150100.1 | - | 3 | 1463 | |||
| AK170298.1 | - | 3 | 1465 | |||
| BC050829.1 | - | 3 | 1447 | |||
| BC110479.1 | - | 1 | 1080 | 40048114 | ||
| BC110480.1 | - | 1 | 1080 | 40048115 | ||
| U02304.1 | - | 2 | 1444 | |||
| X75885.1 | - | 1 | 1089 | |||

Ndst2
Genomic Map