siRNA Details
| Gene Symbol: Foxk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199068.2 | NP_951031.2 | 5 | 1167 | ||
| GenBank mRNA | AK144418.1 | - | 5 | 1060 | ||
| AK147375.1 | - | 5 | 1165 | |||
| BC060238.1 | - | 5 | 1147 | |||
| L26507.1 | - | 5 | 1143 | |||
| U95016.1 | - | 5 | 1143 | |||

Foxk1
Genomic Map