siRNA Details
| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010813.3 | NP_034943.3 | 2 | 688 | ||
| GenBank mRNA | AK147933.1 | - | 2 | 672 | ||
| BC054534.1 | - | 2 | 614 | 5721249 | ||
| U77356.1 | - | 2 | 688 | |||
| Y07609.1 | - | 2 | 634 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mnt| Gene Symbol: Mnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010813.3 | NP_034943.3 | 2 | 688 | ||
| GenBank mRNA | AK147933.1 | - | 2 | 672 | ||
| BC054534.1 | - | 2 | 614 | 5721249 | ||
| U77356.1 | - | 2 | 688 | |||
| Y07609.1 | - | 2 | 634 | |||