siRNA Details
| Gene Symbol: Mobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039364.2 | NP_001034453.1 | 2 | 213 | ||
| NM_001039365.2 | NP_001034454.1 | 3 | 292 | |||
| NM_008614.2 | NP_032640.1 | 3 | 292 | |||
| GenBank mRNA | AK013799.1 | - | 3 | 293 | ||
| AK019323.1 | - | 3 | 293 | |||
| AK028057.1 | - | 3 | 292 | |||
| AK083103.1 | - | 3 | 293 | |||
| AK140055.1 | - | 3 | 293 | |||
| BC048492.1 | - | 3 | 292 | 6593408 | ||
| BY257495.1 | - | 2 | 214 | |||
| U81317.1 | - | 2 | 212 | |||

Mobp
Genomic Map