siRNA Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113374.1 | NP_001106845.1 | 5 | 540 | ||
| NM_001113375.1 | NP_001106846.1 | 5 | 444 | |||
| NM_013826.3 | NP_038854.2 | 6 | 662 | |||
| GenBank mRNA | AF091872.1 | - | 6 | 613 | ||
| AK002719.1 | - | 5 | 508 | |||
| AK010112.1 | - | 6 | 642 | |||
| AK078782.1 | - | 4 | 1304 | |||
| BC004631.1 | - | 5 | 511 | |||
| BC024371.1 | - | 6 | 622 | 4983231 | ||
| CX220128.1 | - | 5 | 394 | |||

Mocs2
Genomic Map