siRNA Details
| Gene Symbol: Mdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008617.2 | NP_032643.2 | 9 | 1015 | ||
| GenBank mRNA | AK002305.1 | - | 9 | 1032 | ||
| AK135162.1 | - | 9 | 1028 | |||
| AK160553.1 | - | 9 | 998 | |||
| AK167809.1 | - | 9 | 998 | |||
| BC023482.1 | - | 9 | 1015 | 5251192 | ||
| DQ402950.1 | - | 8 | 865 | |||
| M16229.1 | - | 9 | 974 | |||

Mdh2
Genomic Map