siRNA Details
| Gene Symbol: Gbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256005.1 | NP_001242934.1 | 11 | 1822 | ||
| NM_008620.3 | NP_032646.2 | 11 | 1759 | |||
| GenBank mRNA | AK220567.1 | - | 11 | 1740 | ||
| BC141364.1 | - | 11 | 1710 | 9055977 | ||
| BC147434.1 | - | 11 | 1710 | 9055976 | ||
| EF494423.1 | - | 11 | 1822 | |||
| EF494424.1 | - | 11 | 1793 | |||

Gbp4
Genomic Map