siRNA Details
| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008625.2 | NP_032651.2 | 27 | 3991 | ||
| GenBank mRNA | BC141338.1 | - | 27 | 3943 | 9055950 | |
| Z11974.1 | - | 27 | 3991 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrc1| Gene Symbol: Mrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008625.2 | NP_032651.2 | 27 | 3991 | ||
| GenBank mRNA | BC141338.1 | - | 27 | 3943 | 9055950 | |
| Z11974.1 | - | 27 | 3991 | |||