siRNA Details
| Gene Symbol: Mrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008626.3 | NP_032652.3 | 20, 21 | 3084 | ||
| GenBank mRNA | BU053305.1 | - | 1, 2 | 146 | ||
| U56734.1 | - | 20, 21 | 3047 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrc2| Gene Symbol: Mrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008626.3 | NP_032652.3 | 20, 21 | 3084 | ||
| GenBank mRNA | BU053305.1 | - | 1, 2 | 146 | ||
| U56734.1 | - | 20, 21 | 3047 | |||