siRNA Details
| Gene Symbol: Mrvi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010826.5 | NP_034956.2 | 14 | 2086 | ||
| NM_194464.3 | NP_919446.2 | 15 | 2329 | |||
| GenBank mRNA | AK141821.1 | - | 10 | 1368 | ||
| AK157169.1 | - | 15 | 1939 | |||
| U63407.1 | - | 15 | 2332 | |||
| U63408.1 | - | 14 | 2081 | |||

Mrvi1
Genomic Map