siRNA Details
| Gene Symbol: Msl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010832.5 | NP_034962.2 | 7 | 781 | ||
| GenBank mRNA | AF117066.2 | - | 7 | 783 | ||
| AK077794.1 | - | 1 | 41 | |||
| AK089368.1 | - | 7 | 864 | |||
| AK146208.1 | - | 7 | 717 | |||
| AK148898.1 | - | 7 | 759 | |||
| BC010226.1 | - | 7 | 742 | 3501126 | ||

Msl3
Genomic Map