siRNA Details
| Gene Symbol: Msl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010832.5 | NP_034962.2 | 7 | 896 | ||
| GenBank mRNA | AF117066.2 | - | 7 | 898 | ||
| AK077794.1 | - | 1 | 156 | |||
| AK089368.1 | - | 7 | 979 | |||
| AK146208.1 | - | 7 | 832 | |||
| AK148898.1 | - | 7 | 874 | |||
| BC010226.1 | - | 7 | 857 | 3501126 | ||

Msl3
Genomic Map