siRNA Details
| Gene Symbol: Mt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008631.2 | NP_032657.1 | 2 | 125 | ||
| GenBank mRNA | AK131894.1 | - | 2 | 125 | ||
| BC099850.1 | - | 2 | 84 | |||
| BC099851.1 | - | 2 | 85 | |||
| BC099852.1 | - | 2 | 85 | |||
| BC099853.1 | - | 2 | 84 | |||
| BC111805.1 | - | 2 | 78 | |||
| BC141870.1 | - | 2 | 86 | 40038882 | ||

Mt4
Genomic Map