siRNA Details
| Gene Symbol: Laptm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008640.2 | NP_032666.2 | 2 | 1073 | ||
| GenBank mRNA | AK013963.1 | - | 1 | 91 | ||
| AK075699.1 | - | 2 | 308 | |||
| AK084515.1 | - | 2 | 716 | |||
| AK088958.1 | - | 2 | 333 | |||
| AK165638.1 | - | 2 | 342 | |||
| AK184706.1 | - | 2 | 117 | |||
| BC086667.1 | - | 2 | 253 | 30633914 | ||
| BC089021.1 | - | 2 | 286 | 6734038 | ||
| U34259.1 | - | 3 | 940 | |||

Laptm4a
Genomic Map