siRNA Details
| Gene Symbol: Fam89b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023166.2 | NP_075655.2 | 2 | 714 | ||
| NM_181452.2 | NP_852117.2 | 2 | 753 | |||
| GenBank mRNA | AB012726.1 | - | 2 | 738 | ||
| AF212920.1 | - | 2 | 735 | |||
| AF212921.1 | - | 2 | 681 | |||
| AK003883.1 | - | 2 | 703 | |||
| AK013256.1 | - | 2 | 682 | |||
| AK181098.1 | - | 2 | 85 | |||
| BC013894.1 | - | 2 | 741 | 4217979 | ||
| BC030163.1 | - | 2 | 724 | 3674980 | ||
| BC056166.1 | - | 2 | 699 | 5715229 | ||

Fam89b
Genomic Map