siRNA Details
| Gene Symbol: Mup3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039544.1 | NP_001034633.1 | 1 | 153 | ||
| GenBank mRNA | AI046324.1 | - | 1 | 211 | ||
| BC019965.1 | - | 1 | 115 | 4194688 | ||
| BC059102.1 | - | 1 | 130 | 4195272 | ||
| BC106100.1 | - | 1 | 119 | 5099753 | ||
| EU882235.1 | - | 1 | 72 | |||
| M27608.1 | - | 1 | 102 | |||
| X03525.1 | - | 1 | 101 | |||

Mup3
Genomic Map