siRNA Details
| Gene Symbol: HTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002159.3 | NP_002150.1 | 5 | 219 | ||
| GenBank mRNA | BC017835.1 | - | 5 | 192 | 4289874 | |
| BP335073.2 | - | 5 | 196 | |||
| CB984538.1 | - | 5 | 223 | |||
| CR627329.1 | - | 3 | 399 | |||
| M26664.1 | - | 4 | 134 | |||

HTN1
Genomic Map