siRNA Details
| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010846.1 | NP_034976.1 | 9 | 1721 | ||
| GenBank mRNA | AK076430.1 | - | 13 | 1473 | ||
| AK150023.1 | - | 8 | 1201 | |||
| AK150236.1 | - | 8 | 1201 | |||
| BC011113.1 | - | 8 | 1706 | 4011821 | ||
| CT010406.1 | - | 1508 | ||||
| EU638327.1 | - | 4 | 526 | |||
| EU638328.1 | - | 7 | 1019 | |||
| M12279.1 | - | 9 | 1721 | |||
| M21038.1 | - | 10 | 1793 | |||
| M21039.1 | - | 10 | 1297 | |||

Mx1
Genomic Map