siRNA Details
| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010846.1 | NP_034976.1 | 1556 | |||
| GenBank mRNA | AK076430.1 | - | 12 | 1308 | ||
| AK150023.1 | - | 7 | 1036 | |||
| AK150236.1 | - | 7 | 1036 | |||
| BC011113.1 | - | 1541 | 4011821 | |||
| CT010406.1 | - | 1343 | ||||
| EU638327.1 | - | 3 | 361 | |||
| EU638328.1 | - | 6 | 854 | |||
| M12279.1 | - | 1556 | ||||
| M21038.1 | - | 1628 | ||||
| M21039.1 | - | 9 | 1132 | |||

Mx1
Genomic Map