siRNA Details
| Gene Symbol: Mybpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008653.2 | NP_032679.2 | 28 | 2988 | ||
| GenBank mRNA | AF097333.1 | - | 28 | 2939 | ||
| AK146661.1 | - | 28 | 2989 | |||
| AK146886.1 | - | 28 | 2989 | |||
| AK169302.1 | - | 27 | 2966 | |||
| AK169306.1 | - | 27 | 2966 | |||
| BC094945.1 | - | 28 | 2985 | |||
| BC131672.1 | - | 28 | 2964 | |||
| BC154408.1 | - | 28 | 3009 | 40110890 | ||

Mybpc3
Genomic Map