siRNA Details
| Gene Symbol: HTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000200.2 | NP_000191.1 | 2, 3 | 172 | ||
| GenBank mRNA | BC009791.1 | - | 2, 3 | 130 | 4293405 | |
| BC095438.1 | - | 2, 3 | 129 | 30352697 | ||
| BP332860.1 | - | 2, 3 | 126 | |||
| CB957931.1 | - | 2, 3 | 172 | |||
| M18372.1 | - | 1, 2 | 92 | |||
| M26665.1 | - | 2, 3 | 126 | |||

HTN3
Genomic Map