siRNA Details
| Gene Symbol: HTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000200.2 | NP_000191.1 | 6 | 349 | ||
| GenBank mRNA | BC009791.1 | - | 6 | 307 | 4293405 | |
| BC095438.1 | - | 6 | 306 | 30352697 | ||
| BP332860.1 | - | 6 | 303 | |||
| CB957931.1 | - | 6 | 349 | |||
| M18372.1 | - | 5 | 269 | |||
| M26665.1 | - | 6 | 303 | |||

HTN3
Genomic Map