siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 10 | 991 | ||
| GenBank mRNA | AK138608.1 | - | 10 | 992 | ||
| BC141362.1 | - | 8 | 908 | 9055974 | ||
| CR756940.1 | - | 4 | 300 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh4| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 10 | 991 | ||
| GenBank mRNA | AK138608.1 | - | 10 | 992 | ||
| BC141362.1 | - | 8 | 908 | 9055974 | ||
| CR756940.1 | - | 4 | 300 | |||