siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 11 | 1066 | ||
| GenBank mRNA | AK138608.1 | - | 11 | 1067 | ||
| BC141362.1 | - | 9 | 983 | 9055974 | ||
| CR756940.1 | - | 5 | 375 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myh4| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 11 | 1066 | ||
| GenBank mRNA | AK138608.1 | - | 11 | 1067 | ||
| BC141362.1 | - | 9 | 983 | 9055974 | ||
| CR756940.1 | - | 5 | 375 | |||