siRNA Details
| Gene Symbol: Myh4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010855.3 | NP_034985.2 | 39 | 5725 | ||
| GenBank mRNA | AJ278733.1 | - | 9 | 1369 | ||
| AJ348869.1 | - | 2 | 140 | |||
| BC029240.1 | - | 6 | 763 | |||
| BC037757.1 | - | 4 | 411 | |||
| BC052786.1 | - | 4 | 419 | |||
| BC141362.1 | - | 37 | 5642 | 9055974 | ||

Myh4
Genomic Map