siRNA Details
| Gene Symbol: Myh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164171.1 | NP_001157643.1 | 22, 23 | 3132 | ||
| NM_010856.4 | NP_034986.1 | 22, 23 | 3068 | |||
| GenBank mRNA | AK142232.1 | - | 22, 23 | 3105 | ||
| BC061145.1 | - | 22, 23 | 3101 | |||
| BC090979.1 | - | 22, 23 | 3101 | |||
| BC110700.1 | - | 22, 23 | 3114 | 6786621 | ||
| BC138027.1 | - | 21, 22 | 3007 | 8861046 | ||
| M76598.1 | - | 20, 21 | 2930 | |||
| M76599.1 | - | 20, 21 | 2930 | |||
| M76600.1 | - | 20, 21 | 2930 | |||
| M76601.1 | - | 22, 23 | 3032 | |||

Myh6
Genomic Map