siRNA Details
| Gene Symbol: Myo7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032394.3 | NP_115770.2 | 15 | 1814 | ||
| GenBank mRNA | AF242411.1 | - | 15 | 1851 | ||
| BC138338.1 | - | 15 | 1814 | 8861358 | ||
| BC138341.1 | - | 15 | 1814 | 8861361 | ||

Myo7b
Genomic Map