siRNA Details
| Gene Symbol: HTR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000864.4 | NP_000855.1 | 1 | 703 | ||
| GenBank mRNA | AF498979.1 | - | 1 | 193 | ||
| BC007720.2 | - | 1 | 703 | 4299633 | ||
| BT007027.1 | - | 1 | 193 | |||
| M81589.1 | - | 1 | 236 | |||
More Information
Set Membership: |
|

HTR1D
Genomic Map