siRNA Details
| Gene Symbol: HTR1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000865.2 | NP_000856.1 | 2 | 1740 | ||
| XM_011535789.2 | XP_011534091.1 | 2 | 2188 | |||
| GenBank mRNA | AF498980.1 | - | 1 | 1037 | ||
| BC069751.1 | - | 2 | 1319 | 7262611 | ||
| M91467.1 | - | 3 | 1603 | |||
More Information
Set Membership: |
|

HTR1E
Genomic Map