siRNA Details
| Gene Symbol: Nap1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008671.2 | NP_032697.2 | 1 | 1540 | ||
| GenBank mRNA | AK051439.1 | - | 1 | 1290 | ||
| AK077430.1 | - | 1 | 1501 | |||
| AK131621.1 | - | 1 | 1418 | |||
| BC115708.1 | - | 1 | 1241 | 40058104 | ||
| BC115761.1 | - | 1 | 1241 | 40058103 | ||
| X92352.1 | - | 1 | 1516 | |||

Nap1l2
Genomic Map