siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010877.4 | NP_035007.1 | 12 | 1274 | ||
GenBank mRNA | AB002664.1 | - | 12 | 1228 | ||
AK036379.1 | - | 12 | 1275 | |||
AK137152.1 | - | 12 | 1329 | |||
AK153447.1 | - | 12 | 1236 | |||
AK155324.1 | - | 12 | 1329 | |||
AK170839.1 | - | 12 | 1299 | |||
AK171059.1 | - | 12 | 1300 | |||
AK171265.1 | - | 12 | 1302 | |||
BC003730.1 | - | 12 | 1222 | 3592460 | ||
BC076609.1 | - | 13 | 2228 | |||
BC094056.1 | - | 12 | 1319 | |||
DQ449939.1 | - | 12 | 1229 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
