siRNA Details
Gene Symbol: Ncf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010877.4 | NP_035007.1 | 3 | 458 | ||
GenBank mRNA | AB002664.1 | - | 3 | 412 | ||
AK036379.1 | - | 3 | 459 | |||
AK137152.1 | - | 3 | 513 | |||
AK153447.1 | - | 3 | 420 | |||
AK155324.1 | - | 3 | 513 | |||
AK170839.1 | - | 3 | 483 | |||
AK170998.1 | - | 3 | 486 | |||
AK171059.1 | - | 3 | 484 | |||
AK171265.1 | - | 3 | 486 | |||
BC003730.1 | - | 3 | 406 | 3592460 | ||
BC076609.1 | - | 4 | 1412 | |||
BC094056.1 | - | 3 | 504 | |||
DQ449939.1 | - | 3 | 413 |
