siRNA Details
| Gene Symbol: Nck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010878.2 | NP_035008.2 | 3 | 640 | ||
| GenBank mRNA | AF043259.1 | - | 3 | 728 | ||
| AF084183.1 | - | 2 | 563 | |||
| AK053155.1 | - | 437 | ||||
| AK054051.1 | - | 2 | 469 | |||
| AK210839.1 | - | 1 | 257 | |||
| BC002015.1 | - | 3 | 640 | 3593441 | ||

Nck1
Genomic Map