siRNA Details
| Gene Symbol: Ncoa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB219976.1 | - | 16, 17 | 3703 | ||
| AF000582.1 | - | 19, 20 | 4064 | |||
| AK035719.1 | - | 6, 7 | 1041 | |||
| BC053387.1 | - | 18, 19 | 4048 | 30054358 | ||
| U39060.1 | - | 19, 20 | 4113 | |||

Ncoa2
Genomic Map