siRNA Details
| Gene Symbol: Ndn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010882.3 | NP_035012.2 | 1 | 869 | ||
| GenBank mRNA | AK005078.1 | - | 1 | 827 | ||
| AK075729.1 | - | 1 | 827 | |||
| AK075735.1 | - | 1 | 815 | |||
| AK076025.1 | - | 1 | 825 | |||
| AK078129.1 | - | 1 | 825 | |||
| AK135444.1 | - | 1 | 825 | |||
| AK149086.1 | - | 1 | 825 | |||
| BC147249.1 | - | 1 | 850 | 9056296 | ||
| BC147250.1 | - | 1 | 850 | 9056298 | ||
| M80840.1 | - | 1 | 805 | |||

Ndn
Genomic Map