siRNA Details
| Gene Symbol: Ndufs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010887.2 | NP_035017.2 | 2, 3 | 219 | ||
| GenBank mRNA | AF124785.1 | - | 2, 3 | 169 | ||
| AK011124.1 | - | 2, 3 | 207 | |||
| AK020552.1 | - | 2, 3 | 175 | |||
| BC004618.1 | - | 2, 3 | 177 | 3585543 | ||
| CJ041886.1 | - | 2, 3 | 385 | |||
| CJ073528.1 | - | 2, 3 | 218 | |||

Ndufs4
Genomic Map