siRNA Details
| Gene Symbol: Neb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010889.1 | NP_035019.1 | 71 | 10804 | ||
| GenBank mRNA | U58108.1 | - | 23 | 4057 | ||
| U58109.1 | - | 6 | 849 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neb| Gene Symbol: Neb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010889.1 | NP_035019.1 | 71 | 10804 | ||
| GenBank mRNA | U58108.1 | - | 23 | 4057 | ||
| U58109.1 | - | 6 | 849 | |||