siRNA Details
| Gene Symbol: Nedd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008682.2 | NP_032708.2 | 7 | 918 | ||
| GenBank mRNA | AK010591.1 | - | 7 | 906 | ||
| AK048393.1 | - | 7 | 870 | |||
| AK085355.1 | - | 7 | 881 | |||
| AK090040.1 | - | 7 | 863 | |||
| AK153746.1 | - | 7 | 920 | |||
| BC052430.1 | - | 7 | 887 | 6836583 | ||
| BC066870.1 | - | 7 | 855 | 30476383 | ||
| D10712.1 | - | 7 | 860 | |||

Nedd1
Genomic Map