siRNA Details
| Gene Symbol: Neo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042752.1 | NP_001036217.1 | 14, 15 | 2607 | ||
| NM_008684.2 | NP_032710.2 | 14, 15 | 2607 | |||
| GenBank mRNA | AK084609.1 | - | 14, 15 | 2596 | ||
| AK087272.1 | - | 11, 12 | 1356 | |||
| BC054540.1 | - | 14, 15 | 2591 | 6406007 | ||
| Y09535.1 | - | 14, 15 | 2474 | |||

Neo1
Genomic Map