siRNA Details
| Gene Symbol: Neurl1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163480.1 | NP_001156952.1 | 3 | 710 | ||
| NM_021360.4 | NP_067335.4 | 3 | 1188 | |||
| GenBank mRNA | AF400063.1 | - | 3 | 759 | ||
| AF401228.1 | - | 3 | 645 | |||
| AK010787.1 | - | 3 | 857 | |||
| AK046382.1 | - | 3 | 987 | |||
| AK078529.1 | - | 1 | 245 | |||
| BC058386.1 | - | 3 | 1013 | 6831952 | ||
| BC099702.1 | - | 3 | 1188 | 6825751 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | Y15160.1 | - | 541 | Chr. - - [-] on Build | ||

Neurl1a
Genomic Map