siRNA Details
| Gene Symbol: PGRMC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006320.4 | NP_006311.2 | 2 | 1517 | ||
| XM_011531533.1 | XP_011529835.1 | 3 | 1720 | |||
| GenBank mRNA | AJ002030.1 | - | 2 | 484 | ||
| AK091741.1 | - | 3 | 454 | |||
| AK094949.1 | - | 1 | 604 | |||
| AK131272.1 | - | 2 | 1517 | |||
| BC016692.1 | - | 2 | 501 | 4067832 | ||
| BC092478.1 | - | 2 | 532 | 4831185 | ||

PGRMC2
Genomic Map